Learn More
Novus Biologicals™ PRMT10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-29238
Product Code. 30045472
  • 571.00€ / 100µg
Description
PRMT10 Polyclonal antibody specifically detects PRMT10 in Human samples. It is validated for Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PRMT10
Polyclonal
Tris-citrate/phosphate buffer, pH 7 to 8
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Immunoprecipitation
Unconjugated
EC 2.1.1.-, FLJ46629, protein arginine methyltransferase 10 (putative), putative protein arginine N-methyltransferase 10
Between 1 and 50
100 μg
Cell Biology
90826
Store at 4°C. Do not freeze.