missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STOML3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584854
This item is not returnable.
View return policy
Description
Immunogen sequence: TIVFPLPMNI LEGIGGVSYD NHKKLPNKA Highest antigen sequence indentity to the following orthologs: Rat - 83%, Mouse - 79%.
STOML3 is required for the function of many mechanoreceptors. It modulates mechanotransduction channels and acid-sensing ion channels (ASIC) proteins. The protein potentiates PIEZO1 and PIEZO2 function by increasing their sensitivity to mechanical stimulations.
Specifications
| STOML3 | |
| Polyclonal | |
| Unconjugated | |
| Stoml3 | |
| Epb7.2l; EPB72-like 3; erythrocyte band 7 integral membrane protein; protein 7.2B; SLP3; SLP-3; sro; stomatin (Epb7.2)-like 3; stomatin (EPB72)-like 3; stomatin like 3; stomatin related olfactory protein SRO; stomatin-like protein 3; Stomatin-related olfactory protein; STOML3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 161003 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TAV4 | |
| Stoml3 | |
| Recombinant protein corresponding to Human STOML3. Recombinant protein control fragment (Product #RP-108251). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction