Learn More
Pro-Lab Diagnostics™ Prolex™ - Blue E. coli 0157 Latex Test Reagent Kits
Product Code. 11969044
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11969044 50 Tests Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11969044 Supplier Pro-Lab Diagnostics™ Supplier No. PL.070B
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Just one colony is sufficient for the test, enabling even limited growth to be detected
  • Developed to remove the possibility of cross-reactions with E. hermanii and other sorbitol negative Escherichia species
  • Latex agglutination
  • Test results in 2 minutes
  • CLIA Complexity: Non-waived
  • Sensitivity of 100%
Specifications
Content And Storage 2°C to 8°C
Clia Complexity Non-waived
Detectable Analytes E. coli Serogroup O157
Sensitivity 1
Type Blue E. coli O157 Latex Test Reagent Kit
Includes E. coli O157 Latex reagent 3.1mL, negative control Latex reagent 1.5mL, positive control 1.5mL, Agglutination cards 25, mixing sticks 75
Quantity 50 Tests
Test Time 2 min.