missing translation for 'onlineSavingsMsg'
Learn More

Pro-Lab Diagnostics™ Prolex™ Streptococcal Grouping Latex Kits

Product Code. 11909044
Change view
Click to view available options
Quantity:
3 mL
Type:
Group A Latex Reagent
Group B Latex Reagent
Group C Latex Reagent
Group D Latex Reagent
Group F Latex Reagent
Group G Latex Reagent
Unit Size:
Pack of 60
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Type unitSize
11909044 3 mL Group A Latex Reagent Pack of 60
11929044 3 mL Group C Latex Reagent Pack of 60
11949044 3 mL Group F Latex Reagent Pack of 60
11939044 3 mL Group D Latex Reagent Pack of 60
11959044 3 mL Group G Latex Reagent Pack of 60
11919044 3 mL Group B Latex Reagent Pack of 60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11909044 Supplier Pro-Lab Diagnostics™ Supplier No. PL.031
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For grouping Lancefield betahaemolytic Streptococci

  • Instant results obtained at room temperature
  • Advanced extraction process using modified nitrous acid reagents
  • Enhanced results with blue carboxylate-modified microparticles (CM-MP)
  • As few as one colony required
  • Colour coded reagents and vials for ease of selection
  • Available individually

Specifications

Color Code Purple
Content And Storage 2°C to 8°C
Type Group A Latex Reagent
Includes 3mL Blue Latex particles coated with purified Rabbit Antibodies
Quantity 3 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.