missing translation for 'onlineSavingsMsg'
Learn More

Proadifen Hydrochloride, Mikromol™

Product Code. 30051999
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051999 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051999 Supplier Mikromol Supplier No. MM3235.00
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Proadifen Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Proadifen Hydrochloride
CAS 62-68-0
Analyte or Component Names Proadifen Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C23 H31 N O2 . Cl H
Certifications/Compliance ISO 17034
Synonym Valeric acid, 2,2-diphenyl-, 2-(diethylamino)ethyl ester hydrochloride (6CI,8CI), 2-(Diethylamino)ethyl 2,2-diphenylvalerate hydrochloride, 2'-Diethylaminoethyl 2,2-diphenylpentanoate hydrochloride, Proadifen hydrochloride, RP 5171, SKF 525, SKF 525A, U 5446, Benzeneacetic acid, alpha-phenyl-alpha-propyl-, 2-(diethylamino)ethyl ester, hydrochloride (1:1), Benzeneacetic acid, alpha-phenyl-alpha-propyl-, 2-(diethylamino)ethyl ester, hydrochloride (9CI), beta-Diethylaminoethyl diphenylpropylacetate hydrochloride
InChI Formula InChI=1S/C23H31NO2.ClH/c1-4-17-23(20-13-9-7-10-14-20,21-15-11-8-12-16-21)22(25)26-19-18-24(5-2)6-3;/h7-16H,4-6,17-19H2,1-3H3;1H
SMILES Cl.CCCC(C(=O)OCCN(CC)CC)(c1ccccc1)c2ccccc2
IUPAC Name 2-(diethylamino)ethyl 2,2-diphenylpentanoate;hydrochloride
Molecular Weight (g/mol) 389.96
Quantity 100 mg
Formula Weight 389.2122 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.