missing translation for 'onlineSavingsMsg'
Learn More

Procainamide Hydrochloride, TRC

Product Code. 30133269
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133269 100 g 100g
30134364 25 g 25g
30011083 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133269 Supplier Toronto Research Chemicals Supplier No. TRCP755130100G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-39-1
Molecular Formula C13 H21 N3 O . Cl H
Molecular Weight (g/mol) 271.79
Synonym Procainamide Hydrochloride, 4-Amino-N-[2-(diethylamino)ethyl]-benzamide hydrochloride (1:1), 4-Amino-N-[2-(diethylamino)ethyl]-benzamide monohydrochloride, p-Amino-N-[2-(diethylamino)ethyl]-benzamide monohydrochloride, 4-Amino-N-[2-(diethylamino)ethyl]benzamide monohydrochloride, Amidoprocaine, Amisalin, Cardiorytmin, HC 5006, Novocaine amide hydrochloride, Novocamid hydrochloride, Procainamide hydrochloride, Procainamide monohydrochloride, Procamid, Procamide, Procan SR, Procapan, Procardyl, Pronestyl, Pronestyl hydrochloride, Spicain amide
IUPAC Name 4-amino-N-[2-(diethylamino)ethyl]benzamide;hydrochloride
SMILES Cl.CCN(CC)CCNC(=O)c1ccc(N)cc1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C13H21N3O.ClH/c1-3-16(4-2)10-9-15-13(17)11-5-7-12(14)8-6-11;/h5-8H,3-4,9-10,14H2,1-2H3,(H,15,17);1H
Formula Weight 271.1451
Percent Purity >95%
Chemical Name or Material Procainamide hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.