Learn More
Procainamide Hydrochloride, TRC
Product Code. 30134364
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134364 25 g 25g
30133269 100 g 100g
30011083 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134364 Supplier Toronto Research Chemicals Supplier No. TRCP75513025G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 614-39-1
Molecular Formula C13 H21 N3 O . Cl H
Molecular Weight (g/mol) 271.79
Synonym Procainamide Hydrochloride, 4-Amino-N-[2-(diethylamino)ethyl]-benzamide hydrochloride (1:1), 4-Amino-N-[2-(diethylamino)ethyl]-benzamide monohydrochloride, p-Amino-N-[2-(diethylamino)ethyl]-benzamide monohydrochloride, 4-Amino-N-[2-(diethylamino)ethyl]benzamide monohydrochloride, Amidoprocaine, Amisalin, Cardiorytmin, HC 5006, Novocaine amide hydrochloride, Novocamid hydrochloride, Procainamide hydrochloride, Procainamide monohydrochloride, Procamid, Procamide, Procan SR, Procapan, Procardyl, Pronestyl, Pronestyl hydrochloride, Spicain amide
IUPAC Name 4-amino-N-[2-(diethylamino)ethyl]benzamide;hydrochloride
SMILES Cl.CCN(CC)CCNC(=O)c1ccc(N)cc1
Specifications
Quantity 25 g
InChI Formula InChI=1S/C13H21N3O.ClH/c1-3-16(4-2)10-9-15-13(17)11-5-7-12(14)8-6-11;/h5-8H,3-4,9-10,14H2,1-2H3,(H,15,17);1H
Formula Weight 271.1451
Percent Purity >95%
Chemical Name or Material Procainamide hydrochloride
Grade HPLC