missing translation for 'onlineSavingsMsg'
Learn More

Prochlorperazine, TRC

Product Code. 30135423
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135423 25 mg 25mg
30136995 50 mg 50mg
17974825 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135423 Supplier Toronto Research Chemicals Supplier No. TRCP75579025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 58-38-8
Molecular Formula C20 H24 Cl N3 S
Molecular Weight (g/mol) 373.94
Synonym 10H-Phenothiazine, 2-chloro-10-[3-(4-methyl-1-piperazinyl)propyl]-, Phenothiazine, 2-chloro-10-[3-(4-methyl-1-piperazinyl)propyl]- (7CI,8CI), 2-Chloro-10-(3-[1-methyl-4-piperazinyl]propyl)phenothiazine, 2-Chloro-10-[3-(4-methyl-1-piperazinyl)propyl]-10H-phenothiazine, 2-Chloro-10-[3-(4-methyl-1-piperazinyl)propyl]phenothiazine, 3-Chloro-10-[3-(1-methyl-4-piperazinyl)propyl]phenothiazine, 6140RP, Bayer A 173, Capazine, Chlormeprazine, Chlorperazine, DDD 66604, Emelent, Kronocin, Meterazin, Meterazine, N-[γ-(4'-Methylpiperazinyl-1')propyl]-3-chlorophenothiazine, NSC 17478, NSC 665801, Nipodal, Proazine, Prochlorpemazine, Prochlorperazin, Prochlorperazine, Prochlorpromazine, RP 6140, SKF 4657, Temetid
IUPAC Name 2-chloro-10-[3-(4-methylpiperazin-1-yl)propyl]phenothiazine
SMILES CN1CCN(CCCN2c3ccccc3Sc4ccc(Cl)cc24)CC1

Tekniske data

Quantity 25 mg
InChI Formula InChI=1S/C20H24ClN3S/c1-22-11-13-23(14-12-22)9-4-10-24-17-5-2-3-6-19(17)25-20-8-7-16(21)15-18(20)24/h2-3,5-8,15H,4,9-14H2,1H3
Formula Weight 373.1379
Percent Purity >95%
Chemical Name or Material Prochlorperazine
Grade HPLC
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.