missing translation for 'onlineSavingsMsg'
Learn More

Procyclidine Hydrochloride, TRC

Product Code. 30138033
Change view
Click to view available options
Quantity:
5 mg
50 mg
1 g
Unit Size:
1g
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138033 50 mg 50mg
30136774 1 g 1g
17985065 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138033 Supplier Toronto Research Chemicals Supplier No. TRCP75584050MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1508-76-5
Molecular Formula C19 H29 N O . Cl H
Molecular Weight (g/mol) 323.9
Synonym 1-Pyrrolidinepropanol, α-cyclohexyl-α-phenyl-, hydrochloride (1:1), 1-Pyrrolidinepropanol, α-cyclohexyl-α-phenyl-, hydrochloride (6CI,7CI,8CI,9CI), (±)-Procyclidine hydrochloride, 1-Cyclohexyl-1-phenyl-3-pyrrolidino-1-propanol hydrochloride, Arpicolin, Kemadrin, Kemadrin hydrochloride, Osnervan, Procyclidine hydrochloride, Tricyclamol hydrochloride
IUPAC Name 1-cyclohexyl-1-phenyl-3-pyrrolidin-1-ylpropan-1-ol;hydrochloride
SMILES Cl.OC(CCN1CCCC1)(C2CCCCC2)c3ccccc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C19H29NO.ClH/c21-19(17-9-3-1-4-10-17,18-11-5-2-6-12-18)13-16-20-14-7-8-15-20;/h1,3-4,9-10,18,21H,2,5-8,11-16H2;1H
Formula Weight 323.2016
Percent Purity >95%
Chemical Name or Material Procyclidine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.