Learn More
behr Labor-Technik™ K 20 Model Standard Kjeldahl Block Digestion System
Complete system with digestion vessel, frame and extraction hood. With 8, 12 or 20 sample positions for standard Kjeldahl digestion vessels with a volume of 250 mL. The block casing as well as the rack for the digestion devices and the hood are made of acid-resistant, non-corrosive stainless steel.
Brand:  behr Labor-Technik™ B00632831
Additional Details : Weight : 32.20000kg
Product Code. 10186394
  • 10620.00€ / Each
Estimated Shipment: 26-06-2024
to see stock.
Add to basket
Récupel-bijdrage uitgesloten
Description
Description
  • High-performance heating and extraction hood with exhaust collector
    • The progress of the digestion can be observed via the removable viewing door on the insert rack
      • behr one-button operation for extremely easy and quick programming
        • Menu navigation in country language
          • 25 freely configurable programs for block temperature and digestion time
            • Storeable applications
              • USB interface
              • Complete system with digestion vessel, frame and extraction hood.
              • Stable and robust design.
Specifications
Specifications
Kjeldahl Block Digestion System
250 mL
430°C
1000 W
20