Learn More
Wheaton™ C Model R2P 2.0 Standard Roller Culture Apparatus
Brand:  Wheaton™ WRBMR5030-C
12654.00 EUR valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 57.60620kg
Product Code. 13453886
  • 13320.00€ / Each
Estimated Shipment: 28-06-2024
to see stock.
Specifications
0.25 to 8.1 rpm
230 V
35 W
50/60 Hz
74 cm
51 kg
For system navigation For Types of Screens
1 Ea.
230 V, 50/60 Hz, 35 W
40°C
UL, CSA, RoHS and CE Standards
63 cm
76 cm
C-Model
80% up to 31°C