missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INPP5B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100575
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total INPP5B.
Cellular calcium signaling is controlled by the production of inositol phosphates (IPs) by phospholipase C in response to extracellular signals. The IP signaling molecules are inactivated by a family of inositol polyphosphate-5-phosphatases (5-phosphatases). INPP5B encodes the type II 5-phosphatase. The protein is localized to the cytosol and mitochondria, and associates with membranes through an isoprenyl modification near the C-terminus. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
Specifications
| INPP5B | |
| Polyclonal | |
| Unconjugated | |
| INPP5B | |
| 5PTase; 75 kDa inositol polyphosphate-5-phosphatase; AW260155; inositol polyphosphate-5-phosphatase B; inositol polyphosphate-5-phosphatase, 75kDa; INPP5B; INPP5P; LOW QUALITY PROTEIN: type II inositol 1,4,5-trisphosphate 5-phosphatase; MGC65156; MGC71303; OCRL2; Phosphoinositide 5-phosphatase; RP11-109P14.7; type II inositol 1,4,5-trisphosphate 5-phosphatase; type II inositol 1,4,5-trisphosphate 5-phosphatase; LOW QUALITY PROTEIN: type II inositol 1,4,5-trisphosphate 5-phosphatase; type II inositol-1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16330, 362590, 3633 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P32019, Q8K337 | |
| INPP5B | |
| A synthesized peptide derived from human INPP5B(Accession P32019), corresponding to amino acid residues R243-K293. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction