Description
- Gastronorm ProfiLine recirculated air freezer
- Steel SwingLine design door
- Fitted lock
- Electronic control system
- External digital temperature display
Specifications
Specifications
| Capacity (Metric) | 1427 L |
| Temperature Range | -10°C to -26°C |
| Color | White |
| Model | GGPV1470X |
| Dimensions (L x W x H) Exterior | 215 H x 143 W x 83 D cm |
| Depth (Metric) Interior | 65 cm |
| Height (Metric) Interior | 155 cm |
| Width (Metric) Interior | 124 cm |
| Voltage | 220/240 V |
| Frequency | 50/60 Hz |
| Show More |