Learn More
WTW™ ProfiLine™ Oxi 3310 Portable Dissolved Oxygen Meters
Product Code. 10176483
Change view
Click to view available options
Type:
Portable DO Meter
Portable DO Meter with Probe
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10176483 Portable DO Meter with Probe Each
10014313 Portable DO Meter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10176483 Supplier WTW™ Supplier No. 2BA301
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
A professional, field-proven DO-meter with backlit LCD graphic display, the ProfiLine Oxygen Meter Oxi 3310 is ideal for mobile measurement combined with data logger and waterproof USB-interface.
Specifications
  • Temperature Range: -5 to 105°C
  • Temperature Accuracy: ±0.1°C
  • Temperature Resolution: 0.1
  • Pressure Range: 500 to 1100mbar
  • Salinity Range: 0 to 70ppt
  • DO Range: 0 to 90mg/L
  • DO Accuracy: ±0.5% of value
  • DO Resolution: 0.01, 0.1mg/L
  • Saturation Range: 0 to 600%
  • Saturation Resolution: 1%
  • Display Type: LCD
  • Probe Type: Galvanic
  • Battery Life: up to 1000 hr.
  • Battery Type: NiMH rechargeable
  • Data Points: 200 manual points, 5000 automatic data sets
  • IP Rating: IP 67
  • Outputs: Waterproof USB
  • Dimensions (D x W x H): 5.7 x 17.7 x 7.9cm
  • Weight: 0.6kg
TRUSTED_SUSTAINABILITY
Specifications
Description Meter with CellOx 325 probe
Includes Meter with carry case including galvanic DO probe CellOx™ 325, cleaning solution, electrolyte, 3 exchange membrane heads, stand, beaker. Including short instruction manual CD-ROM, batteries, driver software for USB, cable
Outputs Waterproof USB
For Use With (Application) For professional field measurements requiring documentation and data transfer
Pressure Range 500 to 1100mbar
Calibration Modes 1
Temperature (Metric) Range -5°C to +105°C
Resolution (Temperature) 0.1
Accuracy (Temperature) ±0.1°C
Display Type LCD
Data Points 200 Manual points, 5000 automatic data sets
Certifications/Compliance cETLus, CE, ISO 9001:2008
IP Rating IP-67
Weight (Metric) 0.6 kg
Warranty 3 Years
Power Supply 4 x 1.5V AA or 4 x 1.2V NiMH batteries
Probe Type Galvanic
Range (DO) 0 to 90mg/L
Range (Salinity) 0 to 70ppt
Resolution (DO) 0.01, 0.1mg/L
Saturation Range 0 to 600%
Saturation Resolution 1%
Battery Life Up to 1000hr.
Battery Type NiMH rechargeable
Type Portable DO Meter with Probe
Width (Metric) 17.7 cm
Height (Metric) 7.9 cm
Depth (Metric) 5.7 cm
Accuracy (DO) ±0.5% of value