Learn More
WTW™ WTW™ ProfiLine™ pH Portable Meters
Product Code. 10136383
Change view
Click to view available options
Type:
pH Meter
pH Meter with Kit
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10136383 pH Meter Each
10703774 pH Meter with Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10136383 Supplier WTW™ Supplier No. 2AA110
  • ONLINE EXCLUSIVE PRICE
    834.00€
    652.00€ / Pack of 1
    Save 182.00€
    21% Off
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Obtain optimum results with this easy and robust portable pH/mV meter with an automatic autoread function for routine pH measurements that is safe, secure, reliable, and prevents errors.
  • Clear keypad with only 6 keys
  • Automatic AutoRead function for repeatable measured values make pH measurement safe and prevent errors
  • Anti-skidding keypad that can be operated with gloves
  • Large display that is clear and easy to read
  • Easy and robust portable pH/mV meter for routine measurement
  • Secure and reliable pH measurement due to repeatable results
  • Waterproof according IP 67, 3-point calibration, integrated calibration timer
TRUSTED_SUSTAINABILITY
Specifications
Includes Short instruction manual, CD-ROM and batteries
Accuracy (Temperature) ±0.1°C
Accuracy (mV) ±0.3, ±1
Accuracy (pH) ±0.1, ±0.01, ±0.005
For Use With (Application) Ideal for all seeking an easy, robust and waterproof instrument for portable pH measuring
Range (pH) -2 to 19.999
Resolution (pH) ±0.005
Range (mV) -1200 to 1200, -2000 to 2000
Resolution (mV) ±0.3, ±1mV
Calibration Points 1, 2 or 3 Calibration WTW Technical Buffer, DIN/NIST buffers
Display Type LCD, 7 Segment
Certifications/Compliance CE
IP Rating IP67
Dimensions (L x W x H) 80 W x 180 H mm
Weight (Metric) 0.6 kg
Warranty 3 Years
Model pH 3110
Power Supply 4 x 1.5V AA or 4 x 1.2V NiMH rechargeable
Resolution All values ±1 digit
Temperature (Metric) Operating -5°C to +105°C
Battery Type AA, NiMH
Type pH Meter
Battery Life 2500 hr.
Dimensions (L x W) 80 W mm
Width (Metric) 80 mm
Height (Metric) 180 mm
Depth (Metric) 55 mm