Learn More
WTW™ ProfiLine™ pH 3310 Portable pH Meter
Product Code. 10186573
Change view
Click to view available options
Type:
Portable pH Meter with Kit
Portable pH meter
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10186573 Portable pH Meter with Kit Each
10564724 Portable pH meter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10186573 Supplier WTW™ Supplier No. 2AA312
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
undefined
Portable, waterproof and robust meters for the determination of the parameters pH, ORP, dissolved oxygen, conductivity and BSB
The pH 3310 is an elegant combination of portable meter and data logger for all those who want to save measuring series automatically and keep processing them on a computer. The meter includes a built-in data memory for 5000 GLP-compliant entries with an interval-controlled logger. All data can conveniently be transmitted to a computer via the USB interface. The CMC function visualizes the optimum measuring range and thus supports correct measurements. The graphic display with clear text menus allows the comfortable and safe operation of the meter.
TRUSTED_SUSTAINABILITY
Specifications
Includes Data logger and USB interface, carry case with pH electrode SenTix™ 41, buffer solution STP 4 and STP 7, stand, beaker, CD-ROM, driver software for USB, batteries, cable
Accuracy (Temperature) ±0.1°C
Accuracy (mV) ±0.3, ±1
Accuracy (pH) ±0.1, ±0.01, ±0.005
For Use With (Application) For professional field measurements requiring documentation and data transfer
Range (pH) -2 to 19.999
Resolution (pH) ±0.005
Range (mV) -1200 to 1200, -2500 to 2500
Resolution (mV) ±0.3, ±1mV
Calibration Points 1, 2, 3, 4, 5 Calibration WTW Technical Buffer, DIN/NIST buffers plus 20 additional buffer set
Display Type LCD
Memory Manual 500/5000 automatic
Certifications/Compliance CE
IP Rating IP66/67
Dimensions (L x W x H) 80 W x 180 H mm
Weight (Metric) 300 g
Warranty 3 Years
Model pH 3310 SET 2
Power Supply 4 x 1.5V AA or 4 x 1.2V NiMH rechargeable
Resolution All values ±1 digit
Temperature (Metric) Operating -5°C to +105°C
Battery Type AA, NiMH
Type Portable pH Meter with Kit
Battery Life 1000 hr.
Dimensions (L x W) 80 W mm
Width (Metric) 80 mm
Height (Metric) 180 mm
Depth (Metric) 55 mm