missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Progesterone-2,2,4,6,6,17alpha,21,21,21-d9, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Progesterone-D9 |
| CAS | 15775-74-3 |
| Molecular Formula | C21 2H9 H21 O2 |
| Quantity | 0.05 g |
| Synonym | Pregn-4-ene-3,20-dione-2,2,4,6,6,17,21,21,21-d9, Progesterone-2,2,4,6,6,17,21,21,21-d9 (8 CI), 2,2,4,6,6,17,21,21,21-d9-Progesterone |
| InChI Formula | InChI=1 S/C21H30O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h12,16-19 H,4-11H2,1-3H3/t16-,17+,18-,19-,20-,21+/m0/s1/i1D3,4D2,8D2,12 D,17 D |
| SMILES | [2 H]C1=C2C([2 H])([2 H])C[C@@H]3 [C@H](CC[C@@]4(C)[C@H]3 CC[C@]4([2 H])C(=O)C([2 H])([2 H])[2 H])[C@@]2(C)CC([2 H])([2 H])C1=O |
| IUPAC Name | (8 S,9 S,10 R,13 S,14 S,17 S)-2,2,4,6,6,17-hexadeuterio-10,13-dimethyl-17-(2,2,2-trideuterioacetyl)-7,8,9,11,12,14,15,16-octahydro-1 H-cyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 323.52 |
| Formula Weight | 323.2811 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction