Learn More
AUTS2 Antibody (CL7067) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76529MFV500
Product Code. 30307073
  • 737.00€ / 0.10mL
Description
AUTS2 Monoclonal antibody specifically detects AUTS2 in Human,Mouse samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AUTS2
Monoclonal
mFluor Violet 500 SE
autism susceptibility candidate 2, autism susceptibility gene 2 protein, autism-related protein 1, KIAA0442FBRSL2, MGC13140
This antibody was developed using a synthetic peptide derived from Q8WXX7, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human, Mouse
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL7067
50mM Sodium Borate
Mouse
Protein A purified
RUO
26053
Store at 4⪚C in the dark.