missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Progranulin/PGRN Antibody (007), HRP , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89675H
This item is not returnable.
View return policy
Description
Progranulin/PGRN Monoclonal antibody specifically detects Progranulin/PGRN in Human samples. It is validated for ELISA
Specifications
| Progranulin/PGRN | |
| Monoclonal | |
| HRP | |
| CLN11, GEP, GP88, granulin precursor, GRN, PCDGF, PEPI, PGRN | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Progranulin/PGRN (NP_002078.1; Met 1-Leu 593). | |
| 0.1 mL | |
| Cancer, Lipid and Metabolism | |
| 2896 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 007 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction