missing translation for 'onlineSavingsMsg'
Learn More

Proguanil Hydrochloride, LoGiCal

Product Code. 30463246
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30463246 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30463246 Supplier LoGiCal Supplier No. LGCFOR0840.00
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlorguanide Hydrochloride
CAS 637-32-1
Analyte or Component Names Chlorguanide Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C11 H16 Cl N5 . Cl H
Certifications/Compliance ISO 9001
Synonym Proguanil hydrochloride, 1-(4-Chlorophenyl)-5-(1-methylethyl)biguanide hydrochloride, Imidodicarbonimidic diamide, N-(4-chlorophenyl)-N'-(1-methylethyl)-, hydrochloride (1:1), Biguanide, 1-(p-chlorophenyl)-5-isopropyl-, monohydrochloride (8CI), Imidodicarbonimidic diamide, N-(4-chlorophenyl)-N'-(1-methylethyl)-, monohydrochloride (9CI), 1-(p-Chlorophenyl)-5-isopropylbiguanide hydrochloride, 3359 RP, Chloroguanide hydrochloride, Chloroquanil, Diguanyl, Drinupal, Drinupal hydrochloride, Guanatol, Guanatol hydrochloride, M 4888, Paludrine, Paludrine hydrochloride, Palusil, Palusil hydrochloride, Proguanil hydrochloride, RP 3359, SN 12,837, SN 12837, Tirian, Tirian hydrochloride
InChI Formula InChI=1S/C11H16ClN5.ClH/c1-7(2)15-10(13)17-11(14)16-9-5-3-8(12)4-6-9;/h3-7H,1-2H3,(H5,13,14,15,16,17);1H
Solution Type Neat
SMILES Cl.CC(C)NC(=N)NC(=N)Nc1ccc(Cl)cc1
IUPAC Name 1-(4-chlorophenyl)-3-(N-propan-2-ylcarbamimidoyl)guanidine;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 290.19
Quantity 10 mg
Formula Weight 289.0861 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.