missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prohibitin Antibody (5H7), Novus Biologicals™
Mouse Monoclonal Antibody
230.00€ - 578.00€
Specifications
| Antigen | Prohibitin |
|---|---|
| Clone | 5H7 |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18150588
|
Novus Biologicals
NBP2-37563 |
0.1 mL |
578.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654366
|
Novus Biologicals
NBP2-37563-0.025ml |
0.025 mL |
230.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Prohibitin Monoclonal specifically detects Prohibitin in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Prohibitin | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse, Rat, Primate, Monkey | |
| PHB1, prohibitin | |
| PHB1 | |
| IgG1 | |
| Unpurified |
| 5H7 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication, Core ESC Like Genes, Ovarian Carcinoma Cell Markers, Stem Cell Markers | |
| Ascites with 0.03% Sodium Azide | |
| 5245 | |
| Purified recombinant fragment of human PHB expressed in E. Coli. | |
| Primary | |
| 30 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title