missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proinsulin Antibody (253627) [DyLight 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB13361E
This item is not returnable.
View return policy
Description
Proinsulin Monoclonal antibody specifically detects Proinsulin in Human, Mouse samples. It is validated for Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Specifications
| Proinsulin | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 253627 | |
| Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| E. coli-derived recombinant human Proinsulin, Phe25-Asn110, Accession # P01308 | |
| 0.1 mL | |
| Diabetes Research, Immune System Diseases, Immunology | |
| 3630 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction