missing translation for 'onlineSavingsMsg'
Learn More

PROK2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA598946

Additional Details : Weight : 0.01000kg

Product Code. 16511772

  • 424.00€ / 100µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Prokineticin-2 (PK2) is a cysteine-rich secreted protein that is expressed in the suprachiasmatic nucleus (SCN) with receptors located in the critical autonomic control centers of the brain. It has a depolarizing effect on neurons expressing the receptor. PK2 is predominantly controlled by the endogenous circadian clock, but light also plays a modulatory role. PK2 functions as a critical SCN output molecule responsible for circadian locomotor rhythms. PK2 expression is high during the day, and responsive to nocturnal light pulses. PK2 also functions as a chemoattractant for subventricular zone-derived neuronal progenitors.
Specifications

Specifications

PROK2
Polyclonal
Unconjugated
PROK2
Bombina variegata 8 kD protein; Bombina variegata 8kDa protein; Bv 8 homolog (Bombina variegata); BV8; Bv8/prokineticin 2-like protein; Bv8/prokineticin 2-like protein splice variant; HH4; homolog of mouse Bv8 (Bombina variegata 8 kDa); homolog of mouse Bv8 (Bombina variegata 8 kDa); prokineticin 2; KAL4; MIT1; PK2; Prok1; Prok2; prokineticin 1; prokineticin 2; prokineticin 2 beta; prokineticin-2; Protein Bv8 homolog; secreted protein
Rabbit
Protein G
RUO
50501
-20°C or -80°C if preferred
Liquid
ELISA, Western Blot
1.5 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q9QXU7
PROK2
Recombinant Mouse Prokineticin-2 protein (27-128AA)
100 μg
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions