missing translation for 'onlineSavingsMsg'
Learn More

Propaphos, TRC

Product Code. 30138689
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138689 250 mg 250mg
30040926 25 mg 25mg
30040530 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138689 Supplier Toronto Research Chemicals Supplier No. TRCP323755250MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7292-16-2
Molecular Formula C13 H21 O4 P S
Molecular Weight (g/mol) 304.34
Synonym Phosphoric acid, 4-(methylthio)phenyl dipropyl ester, Phosphoric acid, p-(methylthio)phenyl dipropyl ester (7CI,8CI), Kayaphos, Kayphosnac, O,O-Dipropyl O-4-methylthiophenyl phosphate, Propaphos
IUPAC Name (4-methylsulfanylphenyl) dipropyl phosphate
SMILES CCCOP(=O)(OCCC)Oc1ccc(SC)cc1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C13H21O4PS/c1-4-10-15-18(14,16-11-5-2)17-12-6-8-13(19-3)9-7-12/h6-9H,4-5,10-11H2,1-3H3
Formula Weight 304.0898
Chemical Name or Material Propaphos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.