missing translation for 'onlineSavingsMsg'
Learn More

Propargyl Bromide (Stablized with MgO), TRC

Product Code. 30411329
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411329 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411329 Supplier Toronto Research Chemicals Supplier No. TRCP76257510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propargyl Bromide
CAS 106-96-7
Molecular Formula C3H3Br
Quantity 10 g
Synonym 3-Bromopropyne, 1-Bromo-2-propyne, 2-Propynyl Bromide, 3-Bromo-1-propyne, 3-Bromopropyne, NSC 8801
InChI Formula InChI=1S/C3H3Br/c1-2-3-4/h1H,3H2
SMILES BrCC#C
IUPAC Name 3-bromoprop-1-yne
Molecular Weight (g/mol) 118.96
Formula Weight 117.9418
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.