missing translation for 'onlineSavingsMsg'
Learn More

Propargyltriphenylphosphonium bromide, 97%, Thermo Scientific™

Product Code. 11475073
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475073 100 g 100g
11455073 5 g 5g
11465073 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475073 Supplier Thermo Scientific Alfa Aesar Supplier No. A12753.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Propargyltriphenylphosphonium bromide can be used in the alcylation of amines. It is a commonly used as a Wittig reagent.

Solubility
Soluble in methanol.

Notes
Hygroscopic. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent and water/moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2091-46-5
Molecular Formula C21H18BrP
Molecular Weight (g/mol) 381.25
MDL Number MFCD00011811
InChI Key AFZDAWIXETXKRE-UHFFFAOYSA-M
Synonym triphenylpropargylphosphonium bromide, propargyltriphenylphosphonium bromide, prop-2-ynyltriphenylphosphonium bromide, triphenyl prop-2-yn-1-yl phosphonium bromide, propargyl triphenylphosphonium bromide, acmc-1cbag, propargyltriphenylphosphoniumbromide, triphenylpropargylphosphoniumbromide, triphenylprop-2-ynylphosphine, bromide, triphenyl-2-propynylphosphonium bromide
PubChem CID 2724214
SMILES [Br-].C#CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 177°C to 182°C
Odor Odorless
Linear Formula HC≡CCH2P(C6H5)3Br
Quantity 100 g
Beilstein 3579775
Sensitivity Hygroscopic
Solubility Information Soluble in methanol.
IUPAC Name triphenyl(prop-2-ynyl)phosphanium;bromide
Formula Weight 381.26
Percent Purity 97%
Chemical Name or Material Propargyltriphenylphosphonium bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.