missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ BAP18 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29049
This item is not returnable.
View return policy
Description
BAP18 Polyclonal antibody specifically detects BAP18 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| BAP18 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| BAP18, BPTF-associated protein of 18 kDa, chromatin complexes subunit BAP18, chromosome 17 open reading frame 49, MGC49942, MLL1/mLL complex subunit C17orf49 | |
| Between 122 and 172 | |
| 100 μg | |
| DNA replication Transcription Translation and Splicing | |
| 124944 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction