missing translation for 'onlineSavingsMsg'
Learn More

Propylhexedrine Hydrochloride, LoGiCal

Product Code. 30458821
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458821 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458821 Supplier LoGiCal Supplier No. LGCFOR0664.00
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propylhexedrine (hydrochloride)
CAS 1007-33-6
Analyte or Component Names Propylhexedrine (hydrochloride)
Shipping Condition Room Temperature
Molecular Formula C10 H21 N . Cl H
Certifications/Compliance ISO 9001
Synonym N,alpha-Dimethyl-cyclohexaneethanamine hydrochloride, N, alpha -dimethyl-cyclohexaneethylamine hydrochloride, (+/-)-Propylhexedrine hydrochloride, Benzedrex hydrochloride, Cyclohexyl(isopropyl)methylammonium chloride, Cyclohexylisopropylmethylamine hydrochloride, Eventin hydrochloride, NSC 170998, NSC 27110, Propylhexedrine hydrochloride, dl-Propylhexedrine hydrochloride, N, alpha -dimethyl-cyclohexaneethylamine hydrochloride
InChI Formula InChI=1S/C10H21N.ClH/c1-9(11-2)8-10-6-4-3-5-7-10;/h9-11H,3-8H2,1-2H3;1H
Solution Type Neat
SMILES Cl.CNC(C)CC1CCCCC1
IUPAC Name 1-cyclohexyl-N-methylpropan-2-amine;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 191.74
Quantity 10 mg
Formula Weight 191.1441 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.