Learn More
Propylthiouracil, TRC
Product Code. 30138557
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138557 10 g 10g
30135568 50 g 50g
17942995 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 30138557 Supplier Toronto Research Chemicals Supplier No. TRCP83831010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 51-52-5
Molecular Formula C7 H10 N2 O S
Molecular Weight (g/mol) 170.23
Synonym Propylthiouracil, 4(1H)-Pyrimidinone, 2,3-dihydro-6-propyl-2-thioxo-, Uracil, 6-propyl-2-thio- (7CI,8CI), 2,3-Dihydro-6-propyl-2-thioxo-4(1H)-pyrimidinone, 2-Mercapto-4-hydroxy-6-n-propylpyrimidine, 2-Mercapto-4-hydroxy-6-propylpyrimidine, 2-Mercapto-6-propylpyrimidin-4-ol, 2-Thio-4-oxo-6-propyl-1,3-pyrimidine, 2-Thio-6-propyl-1,3-pyrimidin-4-one, 6-Propyl-2-thio-2,4(1H,3H)pyrimidinedione, 6-Propyl-2-thiouracil, 6-Propyl-2-thioxo-2,3-dihydro-1H-pyrimidin-4-one, 6-Propylthiouracil, 6-n-Propyl-2-thiouracil, 6-n-Propylthiouracil, NSC 6498, NSC 70461, PTU, PTU (thyreostatic), Procasil, Propacil, Propycil, Propyl-Thiorist, Propyl-Thyracil, Propylthiorit, Prothiucil, Prothiurone, Prothycil, Prothyran, Protiural, Thiuragyl, Thyreostat II
IUPAC Name 6-propyl-2-sulfanylidene-1H-pyrimidin-4-one
SMILES CCCC1=CC(=O)NC(=S)N1
Specifications
Quantity 10 g
InChI Formula InChI=1S/C7H10N2OS/c1-2-3-5-4-6(10)9-7(11)8-5/h4H,2-3H2,1H3,(H2,8,9,10,11)
Formula Weight 170.0514
Percent Purity >95%
Chemical Name or Material 6-Propyl-2-thiouracil
Grade HPLC