missing translation for 'onlineSavingsMsg'
Learn More

Propyphenazone, TRC

Product Code. 30426935
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426935 50 g -
30415341 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30426935 Supplier Toronto Research Chemicals Supplier No. TRCP83838050G
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 479-92-5
Molecular Formula C14 H18 N2 O
Molecular Weight (g/mol) 230.31
Synonym Propyphenazone, 3H-Pyrazol-3-one, 1,2-dihydro-1,5-dimethyl-4-(1-methylethyl)-2-phenyl-, Antipyrine, 4-isopropyl- (6CI,7CI,8CI), 1,2-Dihydro-1,5-dimethyl-4-(1-methylethyl)-2-phenyl-3H-pyrazol-3-one, 1,2-Dihydro-1,5-dimethyl-2-phenyl-4-isopropyl-3H-pyrazol-3-one, 1,5-Dimethyl-2-phenyl-4-propan-2-ylpyrazol-3-one, 1-Phenyl-2,3-dimethyl-4-isopropyl-3-pyrazolin-5-one, 1-Phenyl-2,3-dimethyl-4-isopropylpyrazol-5-one, 4-Isopropyl-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, 4-Isopropylantipyrine, 4-Isopropylphenazone, Arantil P, Budirol, Causyth, Cibalgina, Eufibron, Isopropchin, Isopropylantipyrin, Isopropylantipyrine, Isopropylphenazone, Isopropyrine, Larodon, Propyfenazone, Propyphenazone
IUPAC Name 1,5-dimethyl-2-phenyl-4-propan-2-ylpyrazol-3-one
SMILES CC(C)C1=C(C)N(C)N(C1=O)c2ccccc2

Specifications

Quantity 50 g
InChI Formula InChI=1S/C14H18N2O/c1-10(2)13-11(3)15(4)16(14(13)17)12-8-6-5-7-9-12/h5-10H,1-4H3
Formula Weight 230.1419
Chemical Name or Material Propyphenazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.