Learn More
PROS1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-16910-20UL
 View more versions of this product
Product Code. 18028035
  • 145.00€ / 20µL
Description
The PROS1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PROS1. This antibody recognizes human, mouse antigen. The PROS1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
PROS1
1000 μg/mL
PROS, PROS1, protein S (alpha), PS21, PS22, PS23, PS24, PS25, PSA, Vitamin K dependent protein S
20 μL
5627
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Flow Cytometry (Intracellular Staining)
CoraLite Plus 488
Rabbit
Primary
Human, Mouse
150 kDa