Learn More
PROS1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-16910-20UL
Description
The PROS1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PROS1. This antibody recognizes human, mouse antigen. The PROS1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
| PROS1 | |
| 1000 μg/mL | |
| PROS, PROS1, protein S (alpha), PS21, PS22, PS23, PS24, PS25, PSA, Vitamin K dependent protein S | |
| 20 μL | |
| 5627 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry (Intracellular Staining) | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |