Learn More
DuPont™ Proshield™ 4 Practik
Product Code. 16263512 Shop All DuPont Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 25
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16263512 Medium Pack of 25
16253512 Large Pack of 25
16283502 X-Large Pack of 25
16273502 2X-Large Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 16263512 Supplier DuPont™ Supplier No. D13495537
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Proshield™ 4 Practik suits are the ideal work suits for general maintenance in industrial manufacturing and other harmless environments.
  • Micro-perforated non-woven polyethylene fabric allows unrestricted passage of air and water vapour
  • Certified according to Regulation (EU) 2016/425
  • Chemical protective clothing, Category I
  • Antistatic treatment (EN 1149-5) on inside for comfort
  • Versatile protective garment for non-hazardous substances
TRUSTED_SUSTAINABILITY
Specifications
Size Medium
Gender Unisex
Material Polyethylene
Color White
Disposable Single-use
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper
Sterility Non-sterile
Sleeve Style Elastic