Learn More
Thermo Scientific™ ProSwift™ RP-4H Capillary Monolithic HPLC Columns, Reversed Phase, 100mm length, 0.5mm I.D.
Shop All Thermo Scientific ProductsDescription
Run protein separations at exceptionally high flow rates with Thermo Scientific™ ProSwift™ RP LC Columns. These reversed-phase monolith columns deliver the outstanding resolving power of nonporous analytical media combined with faster separations and analysis than other bead-based columns. ProSwift polymer monolith columns achieve long column lifetime, exhibit reproducibility even after hundreds of runs and have broad pH stability, providing high stringent wash capability (1M NaOH).
Each monolith is a single cylindrical polymer rod containing an uninterrupted, interconnected network of flow-through channels of a specific pore size. These large channels and the monolith's nonporous surfaces provide fast mass transfer for the high-resolution and fast protein separations. The monolith morphology results in low operating backpressure, allowing the use of higher linear velocities with minimal loss of resolution.
Advantages of High Resolution at Exceptionally High Flow Rates
- Highest operational flow rates available
- High throughput and improved productivity
- Excellent stability over a wide pH range of 1 to 14
- Outstanding reproducibility and ruggedness
- High loading capacity
- High speed protein separations
- High sensitivity for LC-MS
- High column-to-column reproducibility
- Wide range of column I.D. and lengths available
- Superior lifetime
- Use Thermo Scientific™ Dionex™ nanoViper™ Fingertight Fittings
- Precolumns are available to preconcentrate and desalt samples.
- You can also use Thermo Scientific™ PepSwift™ Trap Columns for preconcentration and desalting of samples consisting of peptides and proteins without negatively impacting the chromatographic performance or recovery of the compounds.
- Change the selectivity of the separation or improve the trapping efficiency by employing various ion-pairing agents in the loading solvent and/or mobile phases.
Specifications
Specifications
| Column Chemistry | Phenyl |
| Column Format | Capillary Column |
| Diameter (Metric) | 0.5 mm |
| Flow Rate | 12 to 80 μL/min |
| For Use With | LC/MS |
| Length (Metric) | 100 mm |
| Max. Pressure | 2000 psi (140 bar) |
| Particle Size | Monolith |
| pH | 1 to 10 |
| Solvents | Most common Organic Solvents |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.