Learn More
WTW™ Protective Armoring for WTW™ IDS Field Sensor
GREENER_CHOICE
Product Code. 11745214
Change view
Click to view available options
For Use With (Equipment):
IDS FDO 925, TetraCon 925, SensoLyt 900
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11745214 IDS FDO 925, TetraCon 925, SensoLyt 900 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11745214 Supplier WTW™ Supplier No. 903836
In Stock
Specifications
For Use With (Equipment) IDS FDO 925, TetraCon 925, SensoLyt 900
Type Protective Armor
Includes Shroud
Material POM