missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Protein C Polyclonal Antibody

Description
Protein C Polyclonal Antibody for Western Blot
Protein C is a vitamin K-dependent plasma glycoprotein that is cleaved to its activated form by the thrombin-thrombomodulin complex. Protein C in its activated form contains a serine protease domain and functions in degradation of the activated forms of coagulation factors V and VIII. Protein C is a vitamin K-dependent serine protease produced in the liver and made up of 2 polypeptide chains. The 62 kDa proenzyme is activated by thrombin bound to an endothelial surface receptor and the active enzyme cleaves factor Va and VIIIa and thus inhibits blood coagulation. The molecular weight of the active enzyme is 55 kDa and the normal concentrations in human plasma is approximately 1-3 ng/mL because of the very fast turnover, the proenzyme concentration is approximately 3 μg/mL. Mutations in the Protein C gene have been associated with thrombophilia due to protein C deficiency, neonatal purpura fulminans, and recurrent venous thrombosis.
Specifications
Specifications
| Antigen | Protein C |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | PROC |
| Gene Accession No. | P04070 |
| Gene Alias | Activation peptide; Anticoagulant protein C; APC; autoprothrombin IIA; Blood coagulation factor XIV; EDQVDPRLIDGK; inactivator of coagulation factors Va, VIII; PC; prepro-protein C; Proc; PROC1; protein C; protein C (inactivator of coagulation factors Va and VIIIa); protein C prepropeptide; protein C, inactivator of coagulation factors Va and VIIIa; THPH3; THPH4; type I protein C; vitamin K-dependent protein C; Vitamin K-dependent protein C heavy chain; Vitamin K-dependent protein C light chain |
| Gene Symbols | PROC |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?