Learn More
Molecular Probes™ Protein-Protein Crosslinking Kit
Product Code. 10194752
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10194752 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10194752 Supplier Molecular Probes™ Supplier No. P6305
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Protein-Protein Crosslinking Kit
The Protein-Protein Crosslinking Kit provides all of the reagents and purification media required to perform three protein-protein conjugations in which neither protein contains available thiol residues. Briefly, the amines of one of the proteins is thiolated with SPDP and the amines of the second protein are converted to thiol-reactive maleimides with SMCC. This kit was designed to prepare and purify protein-protein conjugates, including crosslinking phycobiliproteins (R-PE) to antibodies, but it can be modified to generate peptide-protein or enzyme-nucleic acid conjugates.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
For Research Use Only. Not for use in diagnostic procedures.