missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Protein S Polyclonal Antibody
GREENER_CHOICE

Product Code. 15995445
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15995445 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15995445 Brand Invitrogen™ Brand No. PA579885

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human HepG2 whole cell.

Protein S is a vitamin K-dependent, potent neutral anticoagulant plasma glycoprotein. Protein S has a molecular weight of approximately 70 kDa and consists of a single polypeptide chain. The concentration of protein S in human plasma is 25 μg/mL, with a half-life of approximately two days. About 40% of Protein S in plasma is in a free form, whereas 60% is complexed with C4b-binding Protein (C4BP). Only the free Protein S functions as a cofactor to antigen presenting cells (APC) in the APC-dependent degradation of Factor Va and Factor VIIIa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Protein S
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene PROS1
Gene Accession No. P07225
Gene Alias AW214361; Preproprotein S; PROS; PROS 1; PROS1; protein S; protein S (alpha); Protein S alpha; protein Sa; proteinS; PS 21; PS 22; PS 23; PS 24; PS 25; PS 26; PS21; PS22; PS23; PS24; PS25; PS26; PSA; rabbit protein S; THPH5; THPH6; unnamed protein product; vitamin K-dependent plasma protein S; vitamin K-dependent protein S; vitamin K-dependent protein S precursor
Gene Symbols PROS1
Host Species Rabbit
Immunogen E. coli-derived human Protein S recombinant protein (Position: L291-S676).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5627
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.