missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Protein S Polyclonal Antibody
Description
Immunogen sequence: KSCEVVSVCL PLNLDTKYEL LYLAEQFAGV VLYLKFRLPE ISRFSAEFDF RTYDSEGVIL YAESIDHSAW LLIALRGGKI EVQLKNEHTS KITTGGDVIN NGLWNMVSVE ELEHSISIKI AKEAVMDINK PGPLFKPENG LLETKV Highest antigen sequence indentity to the following orthologs: Mouse - 79%, Rat - 78%.
Protein S is a vitamin K-dependent, potent neutral anticoagulant plasma glycoprotein. Protein S has a molecular weight of approximately 70 kDa and consists of a single polypeptide chain. The concentration of protein S in human plasma is 25 μg/mL, with a half-life of approximately two days. About 40% of Protein S in plasma is in a free form, whereas 60% is complexed with C4b-binding Protein (C4BP). Only the free Protein S functions as a cofactor to antigen presenting cells (APC) in the APC-dependent degradation of Factor Va and Factor VIIIa.
Specifications
Specifications
| Antigen | Protein S |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | PROS1 |
| Gene Accession No. | P07225 |
| Gene Alias | AW214361; Preproprotein S; PROS; PROS 1; PROS1; protein S; protein S (alpha); Protein S alpha; protein Sa; proteinS; PS 21; PS 22; PS 23; PS 24; PS 25; PS 26; PS21; PS22; PS23; PS24; PS25; PS26; PSA; rabbit protein S; THPH5; THPH6; unnamed protein product; vitamin K-dependent plasma protein S; vitamin K-dependent protein S; vitamin K-dependent protein S precursor |
| Gene Symbols | PROS1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?