missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prox1 Antibody (CL13472) [mFluor Violet 610 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24560MFV610
This item is not returnable.
View return policy
Description
Prox1 Monoclonal antibody specifically detects Prox1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Prox1 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| Homeobox prospero-like protein PROX1, prospero homeobox 1, prospero homeobox protein 1, prospero-related homeobox 1, PROX-1 | |
| This antibody was developed using a recombinant protein derived from Q92786, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Vision | |
| 5629 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL13472 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering