missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ PRP-h5 5 μm 300 Å 4.6 x 150 mm

Product Code. 12610343
Change view
Click to view available options
Length (Metric):
150 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12610343 150 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12610343 Supplier Hamilton™ Supplier No. 79272
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pentafluoro reversed-phase column for unique selectivity. The PRP-h5 utilizes the PRP-3 as its base with a pentafluorinated modification, making it more hydrophobic in nature. The PRP-h5, with its functionality derived from a pentafluorinated polymer bead, delivers a selectivity difference from standard silica C18 stationary phases. This gives chromatographers the desired retention characteristics necessary for certain sample types.

  • Separation Mode – Reversed-Phase
  • Stationary Phase – PRP-h5
  • Format – HPLC Column
  • Hardware Inner Diameter – 4.6 mm
  • Hardware Length – 150 mm
  • Particle Size – 5 μm
  • Packing Material Type – PSDVB
  • Pore Size – 300 Å
  • Hardware Material – Stainless Steel
  • Max Pressure – 5,000 psi
  • Mobile Phase Limits – pH 1-13. 0-100% aqueous, organic modifier
  • Buffer Strength – 0.0-0.5N
  • Temp Limits – 5-85°C
  • Restoration – Run a gradient of 100% water to 100% acetonitrile. Repeat three times
  • Compound Classification – Peptides/Proteins
  • UOM – 1 EACH

Specifications

Column Format HPLC Column
Diameter (Metric) 4.6 mm
Length (Metric) 150 mm
Max. Pressure 5,000 psi
Particle Size 5 μm
pH pH 1-13. 0-100% aqueous, organic modifier
Pore Size 300 Å
Temperature 5° to 85°C
Packing Material PSDVB
Phase Reversed Phase
Material Stainless Steel
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.