missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPF19 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15414-1-AP-20UL
This item is not returnable.
View return policy
Description
PSO4 is the human homolog of yeast Pso4, a gene essential for cell survival and DNA repair.Specifications
| PRPF19 | |
| Polyclonal | |
| Unconjugated | |
| PRPF19 | |
| hPSO4, Nuclear matrix protein 200, Pre mRNA processing factor 19, PRP19, PRP19/PSO4 homolog, PRPF19, PSO4, Senescence evasion factor, SNEV, UBOX4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 246216, 27339, 28000 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99KP6, Q9JMJ4, Q9UMS4 | |
| PRPF19 | |
| PRPF19 Fusion Protein Ag7700 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction