missing translation for 'onlineSavingsMsg'
Learn More

PRR13 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifications

Antigen PRR13
Concentration 0.26 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16892584
View Documents
Proteintech
14101-1-AP-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16882584
View Documents
Proteintech
14101-1-AP-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Negatively regulates TSP1 expression at the level of transcription. This down-regulation was shown to reduce taxane-induced apoptosis.
Specifications

Specifications

PRR13
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Human
Q9NZ81
54458
PRR13 Fusion Protein Ag5280
Primary
-20°C
PRR13
0.26 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
proline rich 13, Proline rich protein 13, PRR13, Taxane resistance protein, TXR1
PRR13
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.