missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRRT1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA563073

Product Code. 15760852

  • 566.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: PAQTAQAPGF VVPTHAGTVG TLPLGGYVAP GYPLQLQPCT AYVPVYPVGT PYAGGTPGGT GVTSTL Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.

PRRT1 is a protein coding gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PRRT1
Polyclonal
Unconjugated
PRRT1
Bat4; Bat-4; C6orf31; C7H6orf31; D17H6S54E; Dispanin subfamily D member 1; DSPD1; G5b; IFITMD7; interferon induced transmembrane protein domain containing 7; Ng5; Orf31; proline rich transmembrane protein 1; proline-rich transmembrane protein 1; PRRT1; prrt1 {ECO:0000250; Synapse differentiation-induced protein 4; SynDIG4; UniProtKB:Q99946}
Rabbit
Antigen affinity chromatography
RUO
80863
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q99946
PRRT1
Recombinant protein corresponding to Human PRRT1. Recombinant protein control fragment (Product #RP-101615).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.