Learn More
PRTN3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25278-1-AP-150UL
 View more versions of this product
Product Code. 16893855
  • 410.00€ / 150µL
Description
Polymorphonuclear leukocyte serine protease that degrades elastin, fibronectin, laminin, vitronectin, and collagen types I, III, and IV (in vitro) and causes emphysema when administered by tracheal insufflation to hamsters.
Specifications
Specifications
PRTN3
Polyclonal
Unconjugated
PRTN3
ACPA, AGP7, C ANCA, C ANCA antigen, Leukocyte proteinase 3, MBN, MBT, Myeloblastin, Neutrophil proteinase 4, P29, PR 3, PR3, proteinase 3, PRTN3, Wegener autoantigen
Rabbit
Antigen Affinity Chromatography
RUO
5657
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P24158
PRTN3
PRTN3 Fusion Protein Ag16765
150 μL