missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSAP Mouse, Clone: SPM312, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 55-MSM3X-P0
This item is not returnable.
View return policy
Specifications
| PSAP | |
| Cytoplasmic | |
| SPM312 | |
| Unconjugated | |
| NP_001127666 | |
| Mouse | |
| 52kDa | |
| Protein A/G Purified | |
| RUO | |
| 55 | |
| IgG1 κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| ACPP | |
| Prostatic acid phosphatase purified from human seminal plasma | |
| PC12 cells or normal prostate or prostate carcinoma | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction