Learn More
Invitrogen™ PSAP Recombinant Rabbit Monoclonal Antibody (24GB5265)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557190
 View more versions of this product
Product Code. 30275749
  • 146.00€ / 20µL
Description
PSAP Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
PSAP is a highly conserved glycoprotein which is a precursor for 4 cleavage products: saposins A, B, C, and D. Each domain of the precursor protein is approximately 80 amino acid residues long with nearly identical placement of cysteine residues and glycosylation sites. Saposins A-D localize primarily to the lysosomal compartment where they facilitate the catabolism of glycosphingolipids with short oligosaccharide groups. The precursor protein exists both as a secretory protein and as an integral membrane protein and has neurotrophic activities.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PSAP
Recombinant Monoclonal
Unconjugated
Psap
A1 activator; AI037048; Cerebroside sulfate activator; Co-beta-glucosidase; Component C; CSAct; Dispersin; GLBA; Glucosylceramidase activator; Proactivator polypeptide; prosaposin; prosaposin (sulfated glycoprotein, sphingolipid hydrolase activator); Protein A; Protein C; Psap; SAP1; SAP-1; SAP-2; Saposin-A; Saposin-B; Saposin-B-Val; Saposin-C; Saposin-D; Sgp1; SGP-1; SGP1A; snoRNA MBII-198; Sphingolipid activator protein 1; Sphingolipid activator protein 2; sphingolipid activator protein-1; sulfated glycoprotein 1; Sulfatide/GM1 activator
Rabbit
Affinity chromatography
RUO
5660
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB5265
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P07602
Psap
A synthesized peptide derived from human PSAP (aa 50-105).