missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
187.00€
Specifications
| Antigen | Proteasome 20S C2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
PSMA1 Polyclonal specifically detects PSMA1 in Human, Mouse, Rat, Canine, Chicken, Chinese Hamster samples. It is validated for Western Blot.Specifications
| Proteasome 20S C2 | |
| Western Blot 1 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Neuroscience, Ubiquitin Proteasome Pathway | |
| PBS with 1 mg/ml BSA with 0.05% Sodium Azide | |
| 5682 | |
| Synthetic Peptide: C P(249) A D E P A E K A D E P M E H(263) | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| 30 kDa |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| EC 3.4.25.1, HC2Proteasome nu chain, Macropain subunit C2, macropain subunit nu, MGC14542, MGC14575, MGC14751, MGC1667, MGC21459, MGC22853, MGC23915, Multicatalytic endopeptidase complex subunit C2, NU30 kDa prosomal protein, PROS-30, PROS30Proteasome component C2, proteasome (prosome, macropain) subunit, alpha type, 1, proteasome subunit alpha type-1, proteasome subunit nu, proteasome subunit, alpha-type, 1, protein P30-33K, PSC2 | |
| PSMA1 | |
| IgG | |
| Affinity Purified | |
| Detects proteasome 20S C2 subunit. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title