Learn More
PSMA7 Monoclonal Antibody (OTI4A9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA810369S
Encompass_Preferred
 View more versions of this product
Product Code. 17553404
  • 242.00€ / 30µL
Description
PSMA7 Monoclonal antibody specifically detects PSMA7 in Human samples. It is validated for Western Blot
Specifications
Specifications
PSMA7
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O14818
Mouse
Affinity chromatography
RUO
5688
-20° C, Avoid Freeze/Thaw Cycles
IgG2a
Western Blot
OTI4A9
Unconjugated
PSMA7
C6; C6-I; epididymis secretory protein Li 276; HEL-S-276; HSPC; proteasome (prosome, macropain) subunit, alpha type 7; proteasome (prosome, macropain) subunit, alpha type, 7; proteasome subunit alpha 4; proteasome subunit alpha 7; proteasome subunit alpha type-7; Proteasome subunit RC6-1; proteasome subunit XAPC7; Psma7; RC6-1; RP5-1005F21.4; testicular tissue protein Li 151; XAPC7
Full length human recombinant protein of human PSMA7 produced in HEK293T cell.