missing translation for 'onlineSavingsMsg'
Learn More

PSMB10 Mouse anti-Human, Porcine, Clone: 1G8C1, Proteintech

Mouse Monoclonal Antibody

145.00€ - 410.00€

Specifications

Antigen PSMB10
Clone 1G8C1
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16898245
View Documents
Proteintech
66403-1-IG-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16888245
View Documents
Proteintech
66403-1-IG-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The 20S proteasome is a protease complex that is responsible for cytosolic protein degradation and generation of peptide ligands for major histocompatabilty complex (MHC) class I molecules, either in their final form or in the form of amino-terminally extended precursors. Upon IFN-gamma stimulation of cells, three constitutively expressed subunits of the 20S proteasome are replaced by inducible subunits LMP2 (low-molecular mass polypeptide 2), LMP7, and MECL-1 (multicatalytic endopeptidase complex-like-1, LMP10). LMP2, LMP7, and MECL-1 subunits form immunoproteasomes, which are associated with more efficient class I antigen processing and presentation. Independent assortment of LMP-2, LMP-7, and MECL-1 into different proteasome complexes can lead to 36 unique proteasome subsets, which may mediate differences in the cleavage specificities/cleavage motifs of proteins subject to constitutive- and immuno-proteasomes.
Specifications

Specifications

PSMB10
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
beta2i, LMP10, Low molecular mass protein 10, Macropain subunit MECl 1, MECL1, Proteasome MECl 1, Proteasome subunit beta 2i, PSMB10
PSMB10
IgG2a
Protein A
Antibody
1G8C1
Western Blot
Unconjugated
Mouse
Pig, Human
P40306
5699, 733626
PSMB10 Fusion Protein Ag8835
Primary
-20°C
PSMB10
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.