missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMG3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15601-1-AP-20UL
This item is not returnable.
View return policy
Description
PSMG3, also named as C7orf48 and PAC3, is a chaperone protein which promotes assembly of the 20S proteasome. It may cooperate with PSMG1-PSMG2 heterodimers to orchestrate the correct assembly of proteasomes.Specifications
| PSMG3 | |
| Polyclonal | |
| Unconjugated | |
| PSMG3 | |
| C7orf48, hPAC3, PAC 3, PAC3, PSMG3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100361517, 66506, 84262 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BT73, Q9CZH3 | |
| Psmg3 | |
| PSMG3 Fusion Protein Ag7984 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction